Assay Details
| Gene Symbol: SLC23A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749803.3 | XP_002749849.1 | 12 | 2466 | 94 | |
| XM_008999514.2 | XP_008997762.1 | 12 | 2493 | 94 | ||
| XM_008999516.2 | XP_008997764.1 | 11 | 2298 | 94 | ||
| XM_008999519.2 | XP_008997767.1 | 11 | 2115 | 94 | ||
More Information
Set Membership: |
|

SLC23A3
Genomic Map