Assay Details
| Gene Symbol: MPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763434.4 | XP_002763480.1 | 11 - 12 | 1389 | 88 | |
| XM_008990125.2 | XP_008988373.1 | 11 - 12 | 1452 | 88 | ||
| GenBank mRNA | EF214922.1 | - | 1 - 2 | 54 | 88 | |
More Information
Set Membership: |
|

MPP1
Genomic Map