Assay Details
| Gene Symbol: CCNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759463.3 | XP_002759509.1 | 11 | 4002 | 113 | |
| GenBank mRNA | EF214993.1 | - | 1 | 126 | 113 | |
More Information
Set Membership: |
|

CCNL1
Genomic Map