Assay Details
| Gene Symbol: LOC100412777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758109.4 | XP_002758155.1 | 9 - 10 | 1089 | 75 | |
| XM_008981310.2 | XP_008979558.1 | 8 - 9 | 939 | 75 | ||
More Information
Set Membership: |
|

LOC100412777
Genomic Map