Assay Details
Gene Symbol: WRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008986697.2 | XP_008984945.1 | 4 - 5 | 485 | 87 | |
XM_008986698.2 | XP_008984946.1 | 3 - 4 | 447 | 87 | ||
XM_008986699.1 | XP_008984947.1 | 3 - 4 | 476 | 87 | ||
XM_008986700.2 | XP_008984948.1 | 3 - 4 | 437 | 87 | ||
XM_008986701.1 | XP_008984949.1 | 4 - 5 | 407 | 87 | ||
XM_008986702.2 | XP_008984950.1 | 3 - 4 | 407 | 87 | ||
XM_008986703.2 | XP_008984951.1 | 3 - 4 | 258 | 87 | ||
XM_017967164.1 | XP_017822653.1 | 4 - 5 | 485 | 87 | ||
XM_017967165.1 | XP_017822654.1 | 3 - 4 | 460 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
