Assay Details
| Gene Symbol: BRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005068.2 | XP_009003316.1 | 2 - 3 | 430 | 118 | |
| XM_009005075.1 | XP_009003323.1 | 2 - 3 | 430 | 118 | ||
| XM_017977245.1 | XP_017832734.1 | 2 - 3 | 1546 | 118 | ||
More Information
Set Membership: |
|

BRD3
Genomic Map