Assay Details
| Gene Symbol: MLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007233.2 | XP_009005481.1 | 7 - 8 | 1157 | 56 | |
| XM_009007235.2 | XP_009005483.1 | 7 - 8 | 788 | 56 | ||
| XM_009007236.2 | XP_009005484.1 | 7 - 8 | 809 | 56 | ||
More Information
Set Membership: |
|

MLC1
Genomic Map