Assay Details
| Gene Symbol: TIMM17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989256.2 | XP_008987504.1 | 2 - 3 | 333 | 64 | |
| XM_008989257.2 | XP_008987505.1 | 2 - 3 | 190 | 64 | ||
| XM_017968472.1 | XP_017823961.1 | 2 - 3 | 185 | 64 | ||
More Information
Set Membership: |
|

TIMM17B
Genomic Map