Assay Details
| Gene Symbol: MED6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754060.3 | XP_002754106.1 | 2 - 3 | 286 | 76 | |
| XM_009006261.2 | XP_009004509.1 | 2 - 3 | 271 | 76 | ||
| XM_017977184.1 | XP_017832673.1 | 2 - 3 | 255 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017977184.1 transcripts.
Set Membership: |
|

MED6
Genomic Map