Assay Details
| Gene Symbol: LOC100405400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007067.2 | XP_009005315.1 | 7 - 8 | 834 | 83 | |
| XM_009007068.2 | XP_009005316.1 | 7 - 8 | 820 | 83 | ||
| XM_009007069.2 | XP_009005317.1 | 7 - 8 | 811 | 83 | ||
More Information
Set Membership: |
|

LOC100405400
Genomic Map