Assay Details
| Gene Symbol: FAM8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746197.3 | XP_002746243.1 | 1 - 2 | 840 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM8A1| Gene Symbol: FAM8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746197.3 | XP_002746243.1 | 1 - 2 | 840 | 77 | |
Set Membership: |
|