Assay Details
| Gene Symbol: CBY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743778.4 | XP_002743824.2 | 2 - 3 | 239 | 102 | |
| XM_008979191.2 | XP_008977439.1 | 3 - 4 | 380 | 102 | ||
| XM_008979198.2 | XP_008977446.1 | 2 - 3 | 241 | 102 | ||
| XM_008979202.2 | XP_008977450.1 | 3 - 4 | 380 | 102 | ||
| XM_008979211.2 | XP_008977459.1 | 3 - 4 | 381 | 102 | ||
| XM_017963747.1 | XP_017819236.1 | 4 - 5 | 472 | 102 | ||
| XM_017963751.1 | XP_017819240.1 | 4 - 5 | 473 | 102 | ||
| XM_017963756.1 | XP_017819245.1 | 3 - 4 | 380 | 102 | ||
| XM_017963759.1 | XP_017819248.1 | 4 - 5 | 472 | 102 | ||
| XM_017963766.1 | XP_017819255.1 | 4 - 5 | 474 | 102 | ||
More Information
Set Membership: |
|

CBY1
Genomic Map