Assay Details
| Gene Symbol: RHEBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752426.4 | XP_002752472.1 | 1 - 2 | 289 | 69 | |
| XM_009003722.2 | XP_009001970.1 | 1 - 2 | 274 | 69 | ||
More Information
Set Membership: |
|

RHEBL1
Genomic Map