Assay Details
| Gene Symbol: KIFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994247.2 | XP_008992495.1 | 6 - 7 | 919 | 68 | |
| XM_008994248.2 | XP_008992496.1 | 6 - 7 | 919 | 68 | ||
| XM_008994249.1 | XP_008992497.1 | 6 - 7 | 957 | 68 | ||
| XM_017971011.1 | XP_017826500.1 | 3 - 4 | 520 | 68 | ||
More Information
Set Membership: |
|

KIFC1
Genomic Map