Assay Details
| Gene Symbol: EEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185044.1 | NP_001171973.1 | 2 - 3 | 304 | 63 | |
| GenBank mRNA | EF059744.1 | - | 2 - 3 | 304 | 63 | |
More Information
Set Membership: |
|

EEF2
Genomic Map