Assay Details
| Gene Symbol: LOC100414136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747891.3 | XP_002747937.1 | 1 - 2 | 153 | 90 | |
| XM_017972267.1 | XP_017827756.1 | 1 - 2 | 153 | 90 | ||
More Information
Set Membership: |
|

LOC100414136
Genomic Map