Assay Details
| Gene Symbol: STX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746968.4 | XP_002747014.1 | 3 - 4 | 410 | 81 | |
| XM_017971400.1 | XP_017826889.1 | 3 - 4 | 1323 | 81 | ||
| XM_017971401.1 | XP_017826890.1 | 3 - 4 | 319 | 81 | ||
| XM_017971402.1 | XP_017826891.1 | 3 - 4 | 410 | 81 | ||
More Information
Set Membership: |
|

STX7
Genomic Map