Assay Details
| Gene Symbol: SLCO4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972159.1 | XP_017827648.1 | 6 - 7 | 2904 | 52 | |
| XM_017972160.1 | XP_017827649.1 | 6 - 7 | 2900 | 52 | ||
More Information
Set Membership: |
|

SLCO4A1
Genomic Map