Assay Details
| Gene Symbol: P2RY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983642.2 | XP_008981890.1 | 1 | 1894 | 72 | |
| XM_017965742.1 | XP_017821231.1 | 1 | 1894 | 72 | ||
More Information
Set Membership: |
|

P2RY1
Genomic Map