Assay Details
| Gene Symbol: ATMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967012.1 | XP_017822501.1 | 1 - 2 | 344 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017967012.1 transcripts.
Set Membership: |
|

ATMIN
Genomic Map