Assay Details
| Gene Symbol: PRIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752618.4 | XP_002752664.1 | 9 - 10 | 1058 | 71 | |
| XM_009004041.2 | XP_009002289.2 | 9 - 10 | 905 | 71 | ||
| XM_017976152.1 | XP_017831641.1 | 9 - 10 | 1058 | 71 | ||
More Information
Set Membership: |
|

PRIM1
Genomic Map