Assay Details
| Gene Symbol: HYAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758381.3 | XP_002758427.1 | 3 - 4 | 1405 | 61 | |
| XM_008981680.2 | XP_008979928.1 | 3 - 4 | 1315 | 61 | ||
More Information
Set Membership: |
|

HYAL2
Genomic Map