Assay Details
| Gene Symbol: GLOD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996316.2 | XP_008994564.1 | 5 - 6 | 631 | 58 | |
| XM_008996317.2 | XP_008994565.1 | 5 - 6 | 659 | 58 | ||
More Information
Set Membership: |
|

GLOD4
Genomic Map