Assay Details
| Gene Symbol: NT5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748722.3 | XP_002748768.1 | 1 - 2 | 259 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NT5C| Gene Symbol: NT5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748722.3 | XP_002748768.1 | 1 - 2 | 259 | 65 | |
Set Membership: |
|