Assay Details
| Gene Symbol: NCR3LG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007750.2 | XP_009005998.1 | 1 - 2 | 150 | 70 | |
| XM_017977937.1 | XP_017833426.1 | 1 - 2 | 150 | 70 | ||
More Information
Set Membership: |
|

NCR3LG1
Genomic Map