Assay Details
| Gene Symbol: PANK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744152.3 | XP_002744198.1 | 6 - 7 | 1364 | 74 | |
| XM_017967563.1 | XP_017823052.1 | 7 - 8 | 1444 | 74 | ||
| XM_017967566.1 | XP_017823055.1 | 7 - 8 | 1194 | 74 | ||
| XM_017967572.1 | XP_017823061.1 | 5 - 6 | 779 | 74 | ||
More Information
Set Membership: |
|

PANK3
Genomic Map