Assay Details
Gene Symbol: ZNF441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008987323.1 | XP_008985571.1 | 3 - 4 | 384 | 93 | |
XM_008987326.1 | XP_008985574.1 | 3 - 4 | 384 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
