Assay Details
| Gene Symbol: TNFSF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205139.2 | NP_001192068.2 | 3 - 4 | 400 | 72 | |
| NM_001278383.1 | NP_001265312.1 | 3 - 4 | 403 | 72 | ||
| GenBank mRNA | HX460499.1 | - | 3 - 4 | 401 | 72 | |
| HX584077.1 | - | 2 - 3 | 153 | 72 | ||
| HX652217.1 | - | 3 - 4 | 401 | 72 | ||

TNFSF12
Genomic Map