Assay Details
| Gene Symbol: EIF2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753141.4 | XP_002753187.3 | 5 - 6 | 802 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF2B1| Gene Symbol: EIF2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753141.4 | XP_002753187.3 | 5 - 6 | 802 | 58 | |
Set Membership: |
|