Assay Details
| Gene Symbol: CAMK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008991712.2 | XP_008989960.1 | 7 - 8 | 1386 | 74 | |
| XM_017969748.1 | XP_017825237.1 | 4 - 5 | 1028 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017969748.1 transcripts.
Set Membership: |
|

CAMK4
Genomic Map