Assay Details
| Gene Symbol: KDELR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744068.4 | XP_002744114.1 | 1 - 2 | 248 | 67 | |
| XM_008983571.2 | XP_008981819.1 | 1 - 2 | 243 | 67 | ||
More Information
Set Membership: |
|

KDELR2
Genomic Map