Assay Details
| Gene Symbol: NMRAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009077.2 | XP_009007325.1 | 4 - 5 | 1583 | 63 | |
| XM_009009078.2 | XP_009007326.1 | 5 - 6 | 874 | 63 | ||
| XM_009009079.1 | XP_009007327.1 | 5 - 6 | 825 | 63 | ||
| XM_009009080.2 | XP_009007328.1 | 5 - 6 | 757 | 63 | ||
| XM_009009081.2 | XP_009007329.1 | 4 - 5 | 648 | 63 | ||
| XM_009009082.2 | XP_009007330.1 | 4 - 5 | 709 | 63 | ||
| XM_009009084.1 | XP_009007332.1 | 4 - 5 | 657 | 63 | ||
| XM_009009087.2 | XP_009007335.1 | 6 - 7 | 899 | 63 | ||
| XM_017978688.1 | XP_017834177.1 | 4 - 5 | 1587 | 63 | ||
| XM_017978689.1 | XP_017834178.1 | 4 - 5 | 665 | 63 | ||
More Information
Set Membership: |
|

NMRAL1
Genomic Map