Assay Details
| Gene Symbol: CAJA-DPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746412.4 | XP_002746458.2 | 1 - 2 | 190 | 89 | |
| XM_008994231.2 | XP_008992479.1 | 1 - 2 | 185 | 89 | ||
| XM_017971018.1 | XP_017826507.1 | 1 - 2 | 215 | 89 | ||
| XM_017971019.1 | XP_017826508.1 | 1 - 2 | 190 | 89 | ||
More Information
Set Membership: |
|

CAJA-DPB1
Genomic Map