Assay Details
| Gene Symbol: FYB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001355.2 | XP_008999603.1 | 13 - 14 | 2117 | 125 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FYB2| Gene Symbol: FYB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001355.2 | XP_008999603.1 | 13 - 14 | 2117 | 125 | |
Set Membership: |
|