Assay Details
| Gene Symbol: ORM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760446.3 | XP_002760492.1 | 4 - 5 | 645 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ORM1| Gene Symbol: ORM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760446.3 | XP_002760492.1 | 4 - 5 | 645 | 100 | |
Set Membership: |
|