Assay Details
| Gene Symbol: H2AFY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756280.4 | XP_002756326.1 | 7 - 8 | 1290 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2AFY2| Gene Symbol: H2AFY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756280.4 | XP_002756326.1 | 7 - 8 | 1290 | 90 | |
Set Membership: |
|