Assay Details
| Gene Symbol: LZTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747309.4 | XP_002747355.2 | 3 - 4 | 795 | 75 | |
| XM_008995666.2 | XP_008993914.1 | 3 - 4 | 795 | 75 | ||
| XM_008995667.2 | XP_008993915.1 | 3 - 4 | 740 | 75 | ||
| XM_008995668.2 | XP_008993916.1 | 3 - 4 | 835 | 75 | ||
More Information
Set Membership: |
|

LZTS3
Genomic Map