Assay Details
| Gene Symbol: LIME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747778.3 | XP_002747824.1 | 4 - 5 | 1271 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LIME1| Gene Symbol: LIME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747778.3 | XP_002747824.1 | 4 - 5 | 1271 | 59 | |
Set Membership: |
|