Assay Details
| Gene Symbol: SDHC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735163.3 | XP_003735211.1 | 3 - 4 | 314 | 68 | |
| XM_017966416.1 | XP_017821905.1 | 3 - 4 | 244 | 68 | ||
| XM_017966418.1 | XP_017821907.1 | 3 - 4 | 180 | 68 | ||
More Information
Set Membership: |
|

SDHC
Genomic Map