Assay Details
| Gene Symbol: FCER1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267729.1 | NP_001254658.1 | 2 - 3 | 95 | 64 | |
| GenBank mRNA | EF534210.1 | - | 2 - 3 | 95 | 64 | |
More Information
Set Membership: |
|

FCER1A
Genomic Map