Assay Details
| Gene Symbol: GABRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744135.3 | XP_002744181.1 | 5 - 6 | 1304 | 79 | |
| XM_002744136.3 | XP_002744182.1 | 5 - 6 | 1304 | 79 | ||
More Information
Set Membership: |
|

GABRG2
Genomic Map