Assay Details
| Gene Symbol: ZNF385B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733158.3 | XP_003733206.1 | 5 - 6 | 936 | 66 | |
| XM_008998610.1 | XP_008996858.1 | 8 - 9 | 1533 | 66 | ||
| XM_008998611.1 | XP_008996859.1 | 7 - 8 | 1494 | 66 | ||
| XM_008998615.2 | XP_008996863.1 | 6 - 7 | 1128 | 66 | ||
| XM_008998616.2 | XP_008996864.1 | 6 - 7 | 2189 | 66 | ||
| XM_008998617.2 | XP_008996865.1 | 6 - 7 | 764 | 66 | ||
| XM_008998618.2 | XP_008996866.1 | 5 - 6 | 1758 | 66 | ||
| XM_008998619.2 | XP_008996867.1 | 5 - 6 | 721 | 66 | ||
More Information
Set Membership: |
|

ZNF385B
Genomic Map