Assay Details
| Gene Symbol: GNPNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753912.4 | XP_002753958.1 | 2 - 3 | 395 | 77 | |
| XM_009006040.2 | XP_009004288.1 | 3 - 4 | 430 | 77 | ||
More Information
Set Membership: |
|

GNPNAT1
Genomic Map