Assay Details
| Gene Symbol: ARL6IP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749434.3 | XP_002749480.1 | 2 - 3 | 1024 | 104 | |
| XM_008998887.2 | XP_008997135.1 | 2 - 3 | 1024 | 104 | ||
| XM_008998889.2 | XP_008997137.1 | 2 - 3 | 1024 | 104 | ||
| XM_017973478.1 | XP_017828967.1 | 2 - 3 | 1024 | 104 | ||
More Information
Set Membership: |
|

ARL6IP6
Genomic Map