Assay Details
| Gene Symbol: TTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757502.2 | XP_002757548.1 | 3 - 4 | 649 | 79 | |
| XM_008980280.1 | XP_008978528.1 | 3 - 4 | 649 | 79 | ||
More Information
Set Membership: |
|

TTL
Genomic Map