Assay Details
| Gene Symbol: AIFM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756279.4 | XP_002756325.1 | 3 - 4 | 1061 | 58 | |
| XM_017979039.1 | XP_017834528.1 | 2 - 3 | 1604 | 58 | ||
More Information
Set Membership: |
|

AIFM2
Genomic Map