Assay Details
| Gene Symbol: FAM207A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986800.2 | XP_008985048.1 | 5 - 6 | 620 | 63 | |
| XM_017967181.1 | XP_017822670.1 | 4 - 5 | 508 | 63 | ||
| XM_017967182.1 | XP_017822671.1 | 4 - 5 | 561 | 63 | ||
More Information
Set Membership: |
|

FAM207A
Genomic Map