Assay Details
| Gene Symbol: TAS1R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750356.3 | XP_002750402.2 | 4 - 5 | 1668 | 53 | |
| XM_017974375.1 | XP_017829864.1 | 7 - 8 | 1792 | 53 | ||
| XM_017974376.1 | XP_017829865.1 | 5 - 6 | 1706 | 53 | ||
More Information
Set Membership: |
|

TAS1R2
Genomic Map