Assay Details
| Gene Symbol: VHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982140.2 | XP_008980388.1 | 2 - 3 | 495 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VHL| Gene Symbol: VHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982140.2 | XP_008980388.1 | 2 - 3 | 495 | 78 | |
Set Membership: |
|