Assay Details
| Gene Symbol: POLR1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746577.4 | XP_002746623.1 | 6 - 7 | 665 | 65 | |
| XM_017971113.1 | XP_017826602.1 | 5 - 6 | 557 | 65 | ||
| XM_017971114.1 | XP_017826603.1 | 6 - 7 | 665 | 65 | ||
| XM_017971115.1 | XP_017826604.1 | 6 - 7 | 665 | 65 | ||
More Information
Set Membership: |
|

POLR1C
Genomic Map